missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARMC6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35622-100ul
This item is not returnable.
View return policy
Description
ARMC6 Polyclonal antibody specifically detects ARMC6 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| ARMC6 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| armadillo repeat containing 6, armadillo repeat-containing protein 6, MGC19595, R30923_1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human ARMC6 (NP_001186125.1).,, Sequence:, MSERCCSRYSSGASIGCTPTSTQAKMVSKRIAQETFDAAVRENIEEFAMGPEEAVKEAVEQFESQGVDLSNIVKTAPKVSADGSQEPTHDILQMLSDLQESVASSRPQEVSAYLTRFCDQCKQDKACRFLAAQKGAYPIIFTAWKLATAGDQGLLLQSLNALSVLTDGQPDLLDAQGLQLLVATLTQNADEADLTCSGIRCVRHACLKHEQNRQDLVKAG | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 93436 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction