missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ArMan
Click to view available options
:
2 mg
Description
ArMan is an aryl mannoside targeting ligand that can be functionalised to the surface of virus like particles (VLPs) via click chemistry. Functionalised particles can be used to target DC-SIGN (dendritic cell-specific intercellular adhesion molecule-3 grabbing nonintegrin) dendritic cells to promote selective coengagement of DC-SIGN and TLR7, resulting in a Th1-type immune response. In vivo, immunization with VLP-ArMan-OvaI/II significantly inhibits tumor growth in a murine melanoma model.
Specifications
Specifications
| Chemical Name or Material | N-(4-(((2R,3S,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)phenethyl)pent-4-ynamide |
| Quantity | 2 mg |
| Target | Buffers, Chelators and Reagents |
| Molecular Formula | C19H25NO7 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction