missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Armadillo 96-well Low Profile Clear PCR Plate, White Wells- PROMO

Product Code. 15311682
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15311682 Clear Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15311682 Supplier Thermo Scientific™ Supplier No. AB3496
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Optically clear skirt—Aids in visualization of liquids and improves stacking capabilities in automated handling systems
  • ANSI:SBS footprint—With rigidity of polycarbonate skirt, the ANSI footprint ensures complete compatibility with robotic systems for high throughput applications
  • Thermostable polycarbonate semi-skirted frame design—Warp-resistant thermal cycling and enhanced mechanical stability in robotic handling
  • Thin walled U-bottom wells—Optimal heat transfer and maximum sample recovery
  • Indented Alphanumeric Grid reference lettering—Integrated frame and well design for improved heat sealing
  • Low profile design—Allows for maximum heat transfer and minimal evaporation when utilizing low volume samples (200 μL working volume). Compatible with Roche LightCycler™ 480 Real-Time PCR systems
  • Available in both clear and white colored wells—Ensures the highest level of sensitivity for all PCR/qPCR reactions
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Array 8 x 12
Color Clear
For Use With (Equipment) BioRad CFX96™, Bio-Rad CFX96 Touch™, Roche LightCycler 480, Eppendorf MasterCycler RealPlex
No. of Wells 96
For Use With (Application) qPCR/Real-Time PCR
Plate Compatibility ANSI footprint ensures complete compatibility with robotic systems for high throughput applications
Quantity 25 Plates
Well Color White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.