missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARL8B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570286
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 100%.
ARL8B may play a role in lysosome motility. may play a role in chromosome segregation.
Specifications
| ARL8B | |
| Polyclonal | |
| Unconjugated | |
| ARL8B | |
| 2610313E07Rik; 3100002J04Rik; ADP ribosylation factor like GTPase 8B; ADP-ribosylation factor like GTPase 8B; ADP-ribosylation factor-like 10C; ADP-ribosylation factor-like 8B; ADP-ribosylation factor-like protein 10C; ADP-ribosylation factor-like protein 8B; Arl10c; Arl8b; Gie1; novel small G protein indispensable for equal chromosome segregation 1; small G protein indispensable for equal chromosome segregation 1; Unknown (protein for MGC:137658) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55207 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NVJ2 | |
| ARL8B | |
| synthetic peptide directed towards the middle region of human ARL8B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction