missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARL8B Polyclonal Antibody
GREENER_CHOICE

Product Code. 16501712
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16501712 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16501712 Supplier Invitrogen™ Supplier No. PA598885

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ARL8B Polyclonal Antibody for Western Blot, IHC (P), ELISA

ARL8B may play a role in lysosome motility. may play a role in chromosome segregation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL8B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ARL8B
Gene Accession No. Q9CQW2, Q9NVJ2
Gene Alias 2610313E07Rik; 3100002J04Rik; ADP ribosylation factor like GTPase 8B; ADP-ribosylation factor like GTPase 8B; ADP-ribosylation factor-like 10C; ADP-ribosylation factor-like 8B; ADP-ribosylation factor-like protein 10C; ADP-ribosylation factor-like protein 8B; Arl10c; Arl8b; Gie1; novel small G protein indispensable for equal chromosome segregation 1; small G protein indispensable for equal chromosome segregation 1; Unknown (protein for MGC:137658)
Gene Symbols ARL8B
Host Species Rabbit
Immunogen Recombinant Human ADP-ribosylation factor-like protein 8B protein (20-186AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55207, 67166
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.