missing translation for 'onlineSavingsMsg'
Learn More

ARL8B Antibody (1A2), Novus Biologicals™

Product Code. 18337629 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18337629 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18337629 Supplier Novus Biologicals Supplier No. H00055207M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ARL8B Monoclonal antibody specifically detects ARL8B in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL8B
Applications ELISA
Classification Monoclonal
Clone 1A2
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH13131
Gene Alias ADP-ribosylation factor-like 10C, ADP-ribosylation factor-like 8B, ADP-ribosylation factor-like protein 10C, ADP-ribosylation factor-like protein 8B, ARL10C, FLJ10702, Gie1, Novel small G protein indispensable for equal chromosome segregation 1
Host Species Mouse
Immunogen ARL8B (AAH13131, 1 a.a. ~ 186 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MLALISRLLDWFRSLFWKEEMELTLVGLQYSGKTTFVNVIASGQFSEDMIPTVGFNMRKVTKGNVTIKIWDIGGQPRFRSMWERYCRGVNAIVYMIDAADREKIEASRNELHNLLDKPQLQGIPVLVLGNKRDLPNALDEKQLIEKMNLSAIQDREICCYSISCKEKDNIDITLQWLIQHSKSRRS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55207
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.