missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ARL8A Recombinant Protein Antigen

Product Code. 18169702 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
This item is not returnable. View return policy

Product Code. 18169702

Brand: Novus Biologicals™ NBP214316PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human ARL8A. The ARL8A Recombinant Protein Antigen is derived from E. coli. The ARL8A Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

This is a blocking peptide for NBP2-14316. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.

TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 127829
Species Human
Purification Method Chromatography
Purity >80%
Concentration 0.5mg/mL
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Blocking/Neutralizing, Control
Gene Symbol ARL8A
Label Type Unlabeled
Molecular Weight (g/mol) 23kDa
Product Type ARL8A
Quantity 0.1 mL
Regulatory Status RUO
Source E.Coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-14316. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Immunogen ALDEKELIEKMNLSAIQDREICCYSISCKEKDNIDITLQWLIQHSKSRRS
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.