missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARL7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100390
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ARL4C.
ADP-ribosylation factor-like 4C is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4C is closely similar to ARL4A and ARL4D and each has a nuclear localization signal and an unusually high guanine nucleotide exchange rate. This protein may play a role in cholesterol transport.
Specifications
| ARL7 | |
| Polyclonal | |
| Unconjugated | |
| ARL4C | |
| ADP ribosylation factor like GTPase 4C; ADP ribosylation factor-like protein 7; ADP-ribosylation factor like GTPase 4C; ADP-ribosylation factor-like 4C; ADP-ribosylation factor-like 7; ADP-ribosylation factor-like protein 4C; ADP-ribosylation factor-like protein 7; ADP-ribosylation factor-like protein LAK; ARL4C; ARL7; LAK | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10123 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P56559 | |
| ARL4C | |
| A synthesized peptide derived from human ARL4C(Accession P56559), corresponding to amino acid residues E148-R192. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction