missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARL6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100129
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total BBS3.
The protein encoded by this gene belongs to the ARF-like (ADP ribosylation factor-like) sub-family of the ARF family of GTP-binding proteins which are involved in regulation of intracellular traffic. Mutations in this gene are associated with Bardet-Biedl syndrome (BBS). Alternative splicing results in multiple transcript variants of this gene. A vision-specific transcript encoding a different protein has been described.
Specifications
| ARL6 | |
| Polyclonal | |
| Unconjugated | |
| ARL6 | |
| 1110018H24Rik; 2210411E14Rik; ADP ribosylation factor like GTPase 6; ADP-ribosylation factor-like 6; ADP-ribosylation factor-like protein 6; ADP-ribosylation-like 6; ADP-ribosylation-like factor homolog ARL6; Arl6; Bardet-Biedl syndrome 3 protein; BBS3; RP55 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 363760, 56297, 84100 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88848, Q9H0F7 | |
| ARL6 | |
| A synthesized peptide derived from human ARL6(Accession Q9H0F7), corresponding to amino acid residues T50-L100. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction