missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARL5B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548169
This item is not returnable.
View return policy
Description
Predicted to react with mouse based on sequence homology.
ARL8 belongs to a family of proteins that are structurally similar to ADP-ribosylation factors. ARLs and ARFs are part of the RAS superfamily of regulatory GTPases.
Specifications
| ARL5B | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q96KC2 | |
| ARL5B | |
| KLH conjugated synthetic peptide between 122-154 amino acids from the C-terminal region of human ARL8. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| ARL5B | |
| 4930587A11Rik; ADP ribosylation factor like GTPase 5B; ADP-ribosylation factor like GTPase 5B; ADP-ribosylation factor-like 5B; ADP-ribosylation factor-like 8; ADP-ribosylation factor-like protein 5B; ADP-ribosylation factor-like protein 8; ADP-ribosylation-like factor 8; ARL5B; Arl8; AW146430 | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 221079 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction