missing translation for 'onlineSavingsMsg'
Learn More

ARL5B Antibody (OTI2D7), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18403495 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403495 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403495 Supplier Novus Biologicals Supplier No. NBP272463MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ARL5B Monoclonal antibody specifically detects ARL5B in Human, Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL5B
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2D7
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias ADP-ribosylation factor-like 5B, ADP-ribosylation factor-like 8, ADP-ribosylation factor-like protein 5B, ADP-ribosylation factor-like protein 8, ADP-ribosylation-like factor 8, ARL8
Host Species Mouse
Immunogen Full length human recombinant protein of human ARL5B(NP_848930) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 221079
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.