missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARL4D Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595838
This item is not returnable.
View return policy
Description
Cellular Location: Golgi apparatus, Lipid-anchor, Membrane Immunogen sequence: RRTDGLVFVV DAAEAERLEE AKVELHRISR ASDNQGVPVL VLANKQDQPG ALSAAEVEKR LAVRELAAAT LTHVQGCSAV DGLGLQQGLE RLYEMILKRK KAARGGKKRR.
ADP-ribosylation factor 4D is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4D is closely similar to ARL4A and ARL4C and each has a nuclear localization signal and an unusually high guanine nucleotide exchange rate. This protein may play a role in membrane-associated intracellular trafficking. Mutations in this gene have been associated with Bardet-Biedl syndrome.
Specifications
| ARL4D | |
| Polyclonal | |
| Unconjugated | |
| ARL4D | |
| 1110036H21Rik; ADP ribosylation factor like GTPase 4D; ADP-ribosylation factor like GTPase 4D; ADP-ribosylation factor-like 4D; ADP-ribosylation factor-like 5; ADP-ribosylation factor-like 6; ADP-ribosylation factor-like protein 4D; ADP-ribosylation factor-like protein 4L; ADP-ribosylation factor-like protein 5; Arf4l; Arfl4; ARL4D; Arl5; ARL6; AW456149; neuroprotective protein 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 303559, 379, 80981 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.36 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P49703, Q99PE9 | |
| ARL4D | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 92-201 of human ARL4D (NP_0016522). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction