missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARL4A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566687
This item is not returnable.
View return policy
Description
Immunogen sequence: MGNGLSDQTSI LSNLPSFQSF HIVILGLDC Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
ARL4A (ADP-ribosylation factor-like 4A) is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4A is similar to ARL4C and ARL4D and each has a nuclear localization signal and an unusually high guaninine nucleotide exchange rate. ARL4A is located in both the nuclear and extranuclear cell compartments. Multiple transcript variants encoding the same protein have been found for this gene.
Specifications
| ARL4A | |
| Polyclonal | |
| Unconjugated | |
| ARL4A | |
| ADP ribosylation factor like GTPase 4A; ADP-ribosylation factor like GTPase 4A; ADP-ribosylation factor-like 4; ADP-ribosylation factor-like 4A; ADP-ribosylation factor-like protein 4A; ADP-ribosylation-like 4; AI467555; Arl4; ARL4A; OTTHUMP00000155200; OTTHUMP00000201575 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10124 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P40617 | |
| ARL4A | |
| Recombinant Human ARL4A. Recombinant protein control fragment (Product #RP-107339). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction