missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARL2BP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101409
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ARL2BP.
ADP-ribosylation factor (ARF)-like proteins (ARLs) comprise a functionally distinct group of the ARF family of RAS-related GTPases. The protein encoded by this gene binds to ARL2. GTP with high affinity but does not interact with ARL2. GDP, activated ARF, or RHO proteins. The lack of detectable membrane association of this protein or ARL2 upon activation of ARL2 is suggestive of actions distinct from those of the ARFs. This protein is considered to be the first ARL2-specific effector identified, due to its interaction with ARL2. GTP but lack of ARL2 GTPase-activating protein activity.
Specifications
| ARL2BP | |
| Polyclonal | |
| Unconjugated | |
| ARL2BP | |
| 1700010P10Rik; 1700027H16Rik; 6330544B05Rik; ADP ribosylation factor like GTPase 2 binding protein; ADP-ribosylation factor like 2 binding protein; ADP-ribosylation factor like GTPase 2 binding protein; ADP-ribosylation factor-like 2 binding protein; ADP-ribosylation factor-like protein 2-binding protein; ADP-ribosylation-like 2 binding protein; AI482273; AI849834; Arf-like 2 binding protein BART1; ARF-like 2-binding protein; ARL2-binding protein; Arl2bp; Bart; BART1; binder of ARF2 protein 1; binder of Arl Two; binder of Arl2; QtsA-10827; retinitis pigmentosa 66 (autosomal recessive); RP66; unnamed protein product | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 107566, 23568, 498910 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q4V8C5, Q9D385, Q9Y2Y0 | |
| ARL2BP | |
| A synthesized peptide derived from human ARL2BP(Accession Q9Y2Y0), corresponding to amino acid residues G106-A156. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction