missing translation for 'onlineSavingsMsg'
Learn More

ARL2BP Antibody (OTI1A10), DyLight 594, Novus Biologicals™

Product Code. 18414338 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414338 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414338 Supplier Novus Biologicals Supplier No. NBP271743DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ARL2BP Monoclonal antibody specifically detects ARL2BP in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL2BP
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1A10
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias ADP-ribosylation factor-like 2 binding protein, ADP-ribosylation factor-like protein 2-binding protein, ARF-like 2-binding protein, BART1binder of Arl Two, BARTArf-like 2 binding protein BART1, Binder of ARF2 protein 1, binder of Arl2
Host Species Mouse
Immunogen Full length recombinant protein of human ARL2BP (NP_036238) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23568
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.