missing translation for 'onlineSavingsMsg'
Learn More

ARL2BP Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18407039 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
18407039 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407039 Supplier Novus Biologicals Supplier No. NBP300281JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ARL2BP Polyclonal antibody specifically detects ARL2BP in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL2BP
Applications ELISA
Classification Polyclonal
Dilution ELISA
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human ARL2BP (Uniprot#: Q9Y2Y0-1; Met1-His163).
Purification Method Antigen and protein A Affinity-purified
Regulatory Status RUO
Target Species Human
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.