missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARL13B Recombinant Rabbit Monoclonal Antibody (HL3092)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556599
This item is not returnable.
View return policy
Description
ARL13B Recombinant Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the ADP-ribosylation factor-like family. The encoded protein is a small GTPase that contains both N-terminal and C-terminal guanine nucleotide-binding motifs. This protein is localized in the cilia and plays a role in cilia formation and in maintenance of cilia. Mutations in this gene are the cause of Joubert syndrome 8. Alternate splicing results in multiple transcript variants.
Specifications
| ARL13B | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q3SXY8 | |
| ARL13B | |
| Recombinant fragment of human ARL13B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL3092 | |
| Unconjugated | |
| ARL13B | |
| A530097K21Rik; A930014M17Rik; ADP ribosylation factor like GTPase 13B; ADP-ribosylation factor like GTPase 13B; ADP-ribosylation factor-like 13B; ADP-ribosylation factor-like 2-like 1; ADP-ribosylation factor-like protein 13B; ADP-ribosylation factor-like protein 2-like 1; Arl13b; Arl2l1; ARL2-like protein 1; C530009C10Rik; hnn; JBTS8; Protein hennin | |
| Rabbit | |
| Protein A | |
| RUO | |
| 200894 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction