missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL13B Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17711-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody detects two specific bands at 60 kDa and 48 kDa. Arl13b is predicted to be a 48 kDa protein, and the 60 kDa band is likely to represent a modified form of Arl13b. ARL13B can be used to mark the cilia.
This gene encodes a member of the ADP-ribosylation factor-like family. The encoded protein is a small GTPase that contains both N-terminal and C-terminal guanine nucleotide-binding motifs. This protein is localized in the cilia and plays a role in cilia formation and in maintece of cilia. Mutations in this gene are the cause of Joubert syndrome 8. Alternate splicing results in multiple transcript variants.Specifications
| ARL13B | |
| Polyclonal | |
| Unconjugated | |
| ARL13B | |
| ARL13B, ARL2 like protein 1, ARL2L1, JBTS8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 200894, 304037, 487934, 68146 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3SXY8, Q640N2 | |
| Arl13b | |
| ARL13B Fusion Protein Ag12015 | |
| 150 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction