missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL13B Mouse anti-Canine, Human, Clone: 1H6C3, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ARL13B |
|---|---|
| Clone | 1H6C3 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16868895
|
Proteintech
66739-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16858895
|
Proteintech
66739-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the ADP-ribosylation factor-like family. The encoded protein is a small GTPase that contains both N-terminal and C-terminal guanine nucleotide-binding motifs. This protein is localized in the cilia and plays a role in cilia formation and in maintece of cilia. Mutations in this gene are the cause of Joubert syndrome 8. Alternate splicing results in multiple transcript variants.Specifications
| ARL13B | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| ARL13B, ARL2 like protein 1, ARL2L1, DKFZp686E2075, DKFZp686L2472, DKFZp686M2074, DKFZp761H079, JBTS8 | |
| ARL13B | |
| IgG2a | |
| Protein A | |
| Antibody |
| 1H6C3 | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Canine | |
| Q3SXY8 | |
| 200894, 487934 | |
| ARL13B Fusion Protein Ag12031 | |
| Primary | |
| -20°C | |
| ARL13B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title