missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ARL10 Polyclonal specifically detects ARL10 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | ARL10 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | ADP-ribosylation factor-like 10, ARL10A |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of human ARL10 |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?