missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ARL 17477 dihydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective neuronal nitrogen oxide synthase (nNOS) inhibitor (IC50 values are 1 and 17 μM for nNOS and endothelial NOS respectively). Reduces ischemic cell damage after middle cerebral artery (MCA) occlusion in rats. Displays a synergistic neuroprotective effect when combined with either an NMDA or AMPA receptor antagonist.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 442.83 |
| Purity | >99% |
| Molecular Weight (g/mol) | 442.827 |
| Chemical Name or Material | ARL 17477 dihydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction