missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Arium™ Tubing Extension Set

Product Code. 17204233 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Arium Smart Station Ultrapure Water
Arium™ Smart Station Pure Water
Unit Size:
1 unit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17204233 Arium™ Smart Station Pure Water 1 unit
17214233 Arium Smart Station Ultrapure Water 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17204233 Supplier Sartorius Supplier No. H2OATESP
  • ONLINE EXCLUSIVE PRICE
    £167.00
    £164.00 / 1 units
    Save £3.00
    1% Off
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Extension set (4 m) for Arium™ Smart Station, used for uncomplicated extension of connection between Arium™ Smart Station and connected water system

  • Extended tubing set used for Smart Station ultrapure and pure for uncomplicated extension of standard supplied tubing set
  • Operating range can be easily increased from 2 to 4 m
TRUSTED_SUSTAINABILITY

Specifications

Color (Overall) White
For Use With (Equipment) Arium™ Smart Station Pure Water
Type Tubing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.