missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Arium™ Smart Station PW

Codice prodotto. 17264233 Sfoglia Tutto Sartorius Prodotti
Cambia vista
Click to view available options
Type:
Remote Dispenser
Remote Dispenser Support
Dimensione della confezione:
1 unit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Type Dimensione della confezione
17264233 Remote Dispenser 1 unit
17254233 Remote Dispenser Support 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17264233 Fornitore Sartorius N. del fornitore H2OARSTPT
  • ONLINE EXCLUSIVE PRICE
    £1825.00
    £1790.00 / 1 units
    Save £35.00
    1% Off
Estimated Shipment: 16-10-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Arium™ Smart Station flexible remote dispenser for pure water

  • Height adjustable display/dispense station for dispensing pure water
  • Compact design for easy and space-saving integration into lab environment
  • Intuitive touch-activated color display with direct access to important functions
  • Flexible and stepless height adjustment to fill different sized containers and beakers
  • Ergonomic design can be optimized for either right or left hand operation
  • Accurate volume dispense for fast and precise buffer and sample prep
  • Adjustable, splash-free flow rate from drop-by-drop to 2 L/min

Equipment supplied:

  • Display/dispense unit with wall bracket
  • Power supply including:
    • IEC connector
    • Connection set

For intended use with:

  • Arium™ Bagtank
  • Arium™ Pro
  • Arium™ Comfort
TRUSTED_SUSTAINABILITY

Specifica

Capacity (Metric) 50 L
Feed Water Monitoring No
Feedwater Source Pure
Flow Rate Up to 2 L/min.
Product Water Type Pure Water
TOC Monitor with UV Intensity Monitoring No
Type Remote Dispenser
Display Touch display
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.