Learn More
Sartorius arium™ Comfort I Combined System (TYPE 1 - Ultrapure Water and TYPE 3 - RO Water)

Shop All Sartorius Products


Description
arium comfort I systems produce ASTM Type 1 ultrapure water and Type 3 pure water. The systems features the latest reverse osmosis technology, and a unique filter cartridge for producing ultrapure water of the highest quality, delivering consistently high-grade water quality at rates of up to 120l/h. The quality of this water even exceeds the ASTM Type 1 specifications. The system's integrated UV lamp (185/254 nm) prevents microbiological growth and reduces the TOC content to a minimum. The current TOC value is continuously checked by an integrated TOC monitor specially developed for this ultrapure water system and is continuously displayed.
- Reliable TOC content of <2 ppb for reproducible results
- Automatically optimised water consumption with iJust
- Easy-to-use - unique glass display with touch function
Benefits:
- Produces Type 3 and Type ASTM 1 water from tap water all in one unit; adapted to the customer's needs; the display will conveniently remain at eye level
- Optimized water consumption
- Meet customer's lab requirements and saves maintenance time
- Safe operation
- Reliable TOC reduction
- Supports reliable recording of important data
- Easy operation
- HPLC
- GC-MS, AAS, ICP-MS
- Ion chromatography
- TOC-Analysis
- Photometry
- Buffer and pH-solutions
- Feed water for ultrapure water systems, rinsing glassware and laboratory vessels, feed water for humidifiers, autoclaves and dish washer
Specifications
Specifications
| Conductivity | 0.055 Product water, Less than1500 feed water |
| Contaminant Rejection (Microbiological) | >99% |
| Contaminant Rejection (Organic) | >99% |
| Bacterial Content | <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge ) |
| Feedwater Connector | 3/8 in. |
| Feed Water Monitoring | No |
| Feedwater Source | Tap |
| Flow Rate | 8 L/hr. |
| Product Water Type | Type 1 to 3 |
| Remote Dispenser | No |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
