missing translation for 'onlineSavingsMsg'
Learn More

Aristolochic acid A, MedChemExpress

Product Code. 16433876
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16433876 25 mg 25mg
16409008 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16433876 Supplier Medchem Express Supplier No. HYN0510
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Aristolochic acid A (Aristolochic acid I; TR 1736) is the main component of plant extract Aristolochic acids, which are found in various herbal plants of genus Aristolochia and Asarum. Aristolochic acid A significantly reduces both activator protein 1 (AP-1) and NF-κB activities. Aristolochic acid A reduces BLCAP gene expression in human cell lines.

Aristolochic acid A (Aristolochic acid I

Specifications

Chemical Name or Material Aristolochic acid A
CAS 313-67-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H11NO7
Quantity 25 mg
Synonym Aristolochic acid I TR 1736
Solubility Information DMSO : 25 mg/mL (73.26 mM; Need ultrasonic)
SMILES OC(C1=CC2=C(OCO2)C3=C4C(C(OC)=CC=C4)=CC([N+]([O-])=O)=C31)=O
Molecular Weight (g/mol) 341.27
Formula Weight 341.27
Percent Purity 98.52%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.