missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aripiprazole, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Aripiprazole
Specifications
Specifications
| Chemical Name or Material | Aripiprazole |
| CAS | 129722-12-9 |
| Analyte or Component Names | Aripiprazole |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H27 Cl2 N3 O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Aripiprazole, 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-2(1H)-quinolinone, 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydrocarbostyril, Abilify, Abilitat, Arpizol, Asprito, OPC 14597, OPC 31, Pripiprazole |
| InChI Formula | InChI=1S/C23H27Cl2N3O2/c24-19-4-3-5-21(23(19)25)28-13-11-27(12-14-28)10-1-2-15-30-18-8-6-17-7-9-22(29)26-20(17)16-18/h3-6,8,16H,1-2,7,9-15H2,(H,26,29) |
| SMILES | Clc1cccc(N2CCN(CCCCOc3ccc4CCC(=O)Nc4c3)CC2)c1Cl |
| IUPAC Name | 7-[4-[4-(2,3-dichlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction