missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aripiprazole metabolite (OPC-3373), Lipomed
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Aripiprazole metabolite (OPC-3373)
Specifications
Specifications
| Chemical Name or Material | 3,4-Dihydro-7-(3'carboxy)propoxy-2(1 H)-quinolinone |
| CAS | 58899-27-7 |
| Analyte or Component Names | 3,4-Dihydro-7-(3'carboxy)propoxy-2(1 H)-quinolinone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H15 N O4 |
| Synonym | Butanoic acid, 4-[(1,2,3,4-tetrahydro-2-oxo-7-quinolinyl)oxy]-, 4-[(1,2,3,4-Tetrahydro-2-oxo-7-quinolinyl)oxy]butanoic acid, 4-[(2-Oxo-3,4-dihydroquinolin-7(1 H)-yl)oxy]butanoic Acid |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | OC(=O)CCCOc1ccc2CCC(=O)Nc2c1 |
| IUPAC Name | 4-[(2-oxo-3,4-dihydro-1 H-quinolin-7-yl)oxy]butanoic acid |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction