missing translation for 'onlineSavingsMsg'
Learn More

Aripiprazole, MedChemExpress

Product Code. 16441736
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
1g
200mg
500mg
This item is not returnable. View return policy

Product Code. 16441736

Brand: Medchem Express HY14546

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Aripiprazole (OPC-14597), an atypical antipsychotic, is a potent and high-affinity dopamine D2 receptor partial agonist. Aripiprazole is an inverse agonist at 5-HT2B and 5-HT2A receptors and displays partial agonist actions at 5-HT1A, 5-HT2C, D3, and D4 receptors. Aripiprazole can be used for the research of schizophrenia and COVID19.

Aripiprazole (OPC-14597), an atypical antipsychotic, is a potent and high-affinity dopamine D2 receptor partial agonist. Aripiprazole is an inverse agonist at 5-HT2B and 5-HT2A receptors and displays partial agonist actions at 5-HT1A, 5-HT2C, D3, and D4 receptors. Aripiprazole can be used for the research of schizophrenia and COVID19.

Specifications

Chemical Name or Material Aripiprazole
CAS 129722-12-9
Color White
Purity Grade Notes Research
Molecular Formula C23H27Cl2N3O2
Quantity 500 mg
Synonym OPC-14597
Solubility Information DMSO : 100 mg/mL (223.02 mM; Need ultrasonic) ∣DMF : 50 mg/mL (111.51 mM; Need ultrasonic)
SMILES O=C1NC2=C(C=CC(OCCCCN3CCN(C4=CC=CC(Cl)=C4Cl)CC3)=C2)CC1
Molecular Weight (g/mol) 448.39
Formula Weight 448.39
Percent Purity 98.39%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.