missing translation for 'onlineSavingsMsg'
Learn More

Aripiprazole, MedChemExpress

Product Code. 16481126
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
1g
200mg
500mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481126 200 mg 200mg
16441736 500 mg 500mg
16406496 100 mg 100mg
16461419 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16481126 Supplier Medchem Express Supplier No. HY14546
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Aripiprazole (OPC-14597), an atypical antipsychotic, is a potent and high-affinity dopamine D2 receptor partial agonist. Aripiprazole is an inverse agonist at 5-HT2B and 5-HT2A receptors and displays partial agonist actions at 5-HT1A, 5-HT2C, D3, and D4 receptors. Aripiprazole can be used for the research of schizophrenia and COVID19.

Aripiprazole (OPC-14597), an atypical antipsychotic, is a potent and high-affinity dopamine D2 receptor partial agonist. Aripiprazole is an inverse agonist at 5-HT2B and 5-HT2A receptors and displays partial agonist actions at 5-HT1A, 5-HT2C, D3, and D4 receptors. Aripiprazole can be used for the research of schizophrenia and COVID19.

Specifications

Chemical Name or Material Aripiprazole
CAS 129722-12-9
Color White
Purity Grade Notes Research
Molecular Formula C23H27Cl2N3O2
Quantity 200 mg
Synonym OPC-14597
Solubility Information DMSO : 100 mg/mL (223.02 mM; Need ultrasonic) ∣DMF : 50 mg/mL (111.51 mM; Need ultrasonic)
SMILES O=C1NC2=C(C=CC(OCCCCN3CCN(C4=CC=CC(Cl)=C4Cl)CC3)=C2)CC1
Molecular Weight (g/mol) 448.39
Formula Weight 448.39
Percent Purity 98.39%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.