missing translation for 'onlineSavingsMsg'
Learn More

Aripiprazole-d8, TRC

Product Code. 30329510
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30329510 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30329510 Supplier Toronto Research Chemicals Supplier No. TRCA77100210MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aripiprazole piperazin-d8
CAS 1089115-06-9
Purity Grade Notes HPLC
Molecular Formula C23 2H8 H19 Cl2 N3 O2
Quantity 10 mg
Synonym Aripiprazole-(piperazin-D8)
InChI Formula InChI=1S/C23H27Cl2N3O2/c24-19-4-3-5-21(23(19)25)28-13-11-27(12-14-28)10-1-2-15-30-18-8-6-17-7-9-22(29)26-20(17)16-18/h3-6,8,16H,1-2,7,9-15H2,(H,26,29)/i11D2,12D2,13D2,14D2
SMILES [2H]C1([2H])N(CCCCOc2ccc3CCC(=O)Nc3c2)C([2H])([2H])C([2H])([2H])N(c4cccc(Cl)c4Cl)C1([2H])[2H]
IUPAC Name 7-[4-[2,2,3,3,5,5,6,6-octadeuterio-4-(2,3-dichlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one
Molecular Weight (g/mol) 456.43
Formula Weight 455.1982
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.