missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aripiprazole, 1mg/ml in Acetonitrile, Lipomed
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Aripiprazole, 1mg/ml in Acetonitrile
Specifications
Specifications
| Chemical Name or Material | Aripiprazole |
| CAS | 129722-12-9 |
| Analyte or Component Names | Aripiprazole |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H27 Cl2 N3 O2 |
| Concentration | 1 mg/mL |
| Synonym | Aripiprazole, 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-2(1 H)-quinolinone, 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydrocarbostyril, Abilify, Abilitat, Arpizol, Asprito, OPC 14597, OPC 31, Pripiprazole |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Clc1cccc(N2CCN(CCCCOc3ccc4CCC(=O)Nc4c3)CC2)c1Cl |
| IUPAC Name | 7-[4-[4-(2,3-dichlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydro-1 H-quinolin-2-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction