missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARIH2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17247643 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17247643 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17247643 Supplier Invitrogen™ Supplier No. PA5114879

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ARIH2.

E3 ubiquitin-protein ligase mediating 'Lys-48'-and 'Lys-63'-linked polyubiquitination and subsequent proteasomal degradation of modified proteins. May play a role in myelopoiesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARIH2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Arih2
Gene Accession No. O95376, Q9Z1K6
Gene Alias AI843547; all-trans retinoic acid inducible RING finger; ARI2; ARI-2; ariadne 2; ariadne homolog 2; ariadne homolog 2 (Drosophila); ariadne RBR E3 ubiquitin protein ligase 2; ARIH2; E3 ubiquitin-protein ligase ARIH2; FLJ10938; FLJ33921; HT005; OTTHUMP00000210388; protein ariadne-2 homolog; Protein ariadne-2-like protein-like protein; RING-type E3 ubiquitin transferase ARIH2; Triad1; Triad1 protein; UbcM4-interacting protein 48; Uip48
Gene Symbols Arih2
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARIH2(Accession O95376), corresponding to amino acid residues N311-A361.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10425, 23807, 316005
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.