missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARIH2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15460
This item is not returnable.
View return policy
Description
ARIH2 Polyclonal specifically detects ARIH2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| ARIH2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| ARI-2, ariadne (Drosophila) homolog 2, ariadne homolog 2 (Drosophila), E3 ubiquitin-protein ligase ARIH2, EC 6.3.2.-, FLJ10938, FLJ33921, Protein ariadne-2 homolog, Triad1 protein, TRIAD1ARI2all-trans retinoic acid inducible RING finger | |
| Rabbit | |
| 58 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| 0.85mg/mL | |
| Western Blot 1:500-1:3000 | |
| O95376 | |
| ARIH2 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human ARIH2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 10425 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction