Learn More
Invitrogen™ ARID5A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113937
Description
Immunogen sequence: LWKNVYDELG GSPGSTSAAT CTRRHYERLV LPYVRHLKGE DDKPLPTSKP For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Members of the ARID protein family, including ARID5A, have diverse functions but all appear to play important roles in development, tissue-specific gene expression, and regulation of cell growth (Patsialou et al., 2005 [PubMed 15640446]).
Specifications
| ARID5A | |
| Polyclonal | |
| Unconjugated | |
| ARID5A | |
| ARID domain-containing protein 5A; Arid5a; AT rich interactive domain 5A (Mrf1 like); AT rich interactive domain 5A (MRF1-like); AT-rich interaction domain 5A; AT-rich interactive domain-containing protein 5A; D430024K22Rik; hypothetical protein LOC524118; LOW QUALITY PROTEIN: AT-rich interactive domain-containing protein 5A; modulator recognition factor 1; modulator recognition factor I; MRF1; MRF-1; RFVG5814; RP11-363D14 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10865 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q03989 | |
| ARID5A | |
| Synthetic peptide directed towards the N terminal region of human ARID5A (aa 110-159). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.