missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARID4B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24499-1-AP-150UL
This item is not returnable.
View return policy
Description
The exact function of C18orf25 remains unknown. There are two named isoforms.Specifications
| ARID4B | |
| Polyclonal | |
| Unconjugated | |
| ARID4B | |
| ARID4B, BRCAA1, RBBP1L1, RBP1L1, SAP180 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51742 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q4LE39 | |
| ARID4B | |
| ARID4B Fusion Protein Ag21462 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction