missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARID4B Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67384-1-IG-20UL
This item is not returnable.
View return policy
Description
The ARID4B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ARID4B. This antibody recognizes human antigen. The ARID4B antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| ARID4B | |
| Monoclonal | |
| 1600 μg/mL | |
| BCAA; BRCAA1; DKFZp313M2420; MGC163290; RBBP1L1; RBP1L1; SAP180 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| Monoclonal Antibody | |
| IgG1 |
| Western Blot, ELISA | |
| 1H3E2 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 51742 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction