missing translation for 'onlineSavingsMsg'
Learn More

ARID3B Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18418374 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418374 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418374 Supplier Novus Biologicals Supplier No. NBP130457AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ARID3B Polyclonal antibody specifically detects ARID3B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARID3B
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ARID domain-containing protein 3B, AT rich interactive domain 3B (BRIGHT- like), AT rich interactive domain 3B (BRIGHT-like), AT-rich interactive domain-containing protein 3B, BDP, bright and dead ringer protein, DRIL2
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 511 and 561 of human AT rich interactive domain 3B using the numbering given in entry EAW99328.1 (GeneID 10620).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10620
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.