missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARID3B Antibody (008), DyLight 550, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89222R
This item is not returnable.
View return policy
Description
ARID3B Monoclonal specifically detects ARID3B in Human samples. It is validated for Western Blot.
Specifications
| ARID3B | |
| Monoclonal | |
| DyLight 550 | |
| ARID domain-containing protein 3B, AT rich interactive domain 3B (BRIGHT- like), AT rich interactive domain 3B (BRIGHT-like), AT-rich interactive domain-containing protein 3B, BDP, bright and dead ringer protein, DRIL2 | |
| This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human ARID3B. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 8 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 10620 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction