missing translation for 'onlineSavingsMsg'
Learn More

ARID3B Antibody (008), DyLight 488, Novus Biologicals™

Product Code. 18386261 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386261 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386261 Supplier Novus Biologicals Supplier No. NBP289222G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ARID3B Monoclonal specifically detects ARID3B in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARID3B
Applications Western Blot
Classification Monoclonal
Clone 8
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias ARID domain-containing protein 3B, AT rich interactive domain 3B (BRIGHT- like), AT rich interactive domain 3B (BRIGHT-like), AT-rich interactive domain-containing protein 3B, BDP, bright and dead ringer protein, DRIL2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human ARID3B.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10620
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.