missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ARID2 Polyclonal antibody specifically detects ARID2 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ARID2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ARID domain-containing protein 2, AT rich interactive domain 2 (ARID, RFX-like), BAF200KIAA1557AT-rich interactive domain-containing protein 2, BRG1-associated factor 200, DKFZp686G052, DKFZp779P0222, FLJ30619, p200, Zinc finger protein with activation potential, zipzap, zipzap/p200 |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 1600 and 1650 of human AT rich interactive domain 2 using the numbering given in entry NP_689854.2 (GeneID 196528). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?