missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARID1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553438
This item is not returnable.
View return policy
Description
Immunogen sequence: SQYGPQGNYS RPPAYSGVPS ASYSGPGPGM GISANNQMHG QGPSQPCGAV PLGRMPSAGM QNRPFPGNMS SMTPSSPGMS QQGGPGMGPP MPTVNRKAQE AAAAVMQAAA NSAQSR Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.
This locus encodes an AT-rich DNA interacting domain-containing protein. The encoded protein is a component of the SWI/SNF chromatin remodeling complex and may play a role in cell-cycle activation. The protein encoded by this locus is similar to AT-rich interactive domain-containing protein 1A. These two proteins function as alternative, mutually exclusive ARID-subunits of the SWI/SNF complex. The associated complexes play opposing roles. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq, Feb 2012].
Specifications
| ARID1B | |
| Polyclonal | |
| Unconjugated | |
| ARID1B | |
| 6A3-5; 8030481M12; 9330189K18Rik; AI836955; Ardi1b; ARID domain-containing protein 1B; ARID1B; AT rich interactive domain 1B (Swi1 like); AT rich interactive domain 1B (SWI1-like); AT rich interactive domain 1B (SWI-like); AT-rich interaction domain 1B; AT-rich interactive domain-containing protein 1B; B230217J03Rik; BAF250B; BRG1-associated factor 250b; BRG1-binding protein ELD/OSA1; BRG1-binding protein hELD/OSA1; BRIGHT; CSS1; DAN15; ELD (eyelid)/OSA protein; ELD/OSA1; hOsa2; KIAA1235; mKIAA1235; MRD12; Osa homolog 2; OSA2; P250R; Smcf1; smcf-1; transcription factor 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57492 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NFD5 | |
| ARID1B | |
| Recombinant protein corresponding to Human ARID1B. Recombinant protein control fragment (Product #RP-91593). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction