missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARID1A Antibody (CL3595) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61623MFV450
This item is not returnable.
View return policy
Description
ARID1A Monoclonal antibody specifically detects ARID1A in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| ARID1A | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| ARID domain-containing protein 1A, AT rich interactive domain 1A (SWI- like), AT rich interactive domain 1A (SWI-like), AT-rich interactive domain-containing protein 1A, B120SWI-like protein, BAF250a, BAF250SWI/SNF complex protein p270, BM029, brain protein 120, BRG1-associated factor 250, BRG1-associated factor 250a, C10rf4, C1orf4SWI/SNF-related, matrix-associated, actin-dependent regulator of chromatinsubfamily F member 1, chromatin remodeling factor p250, hELD, hOSA1, matrix associated, actin dependent regulator of chromatin, Osa homolog 1, OSA1, OSA1 nuclear protein, P270, SMARCF1, subfamily f, member 1 | |
| This antibody was developed using a recombinant protein derived from O14497, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Signal Transduction | |
| 8289 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL3595 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction