missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGEF9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20042-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to ARHGEF9.
The protein encoded by this gene is a Rho-like GTPase that switches between the active state and inactive state to regulate CDC42 and other genes. Defects in this gene are a cause of startle disease with epilepsy, also known as hyperekplexia with epilepsy. Three transcript variants encoding different isoforms have been found for this gene.Specifications
| ARHGEF9 | |
| Polyclonal | |
| Unconjugated | |
| ARHGEF9 | |
| ARHGEF9, COLLYBISTIN, HPEM 2, KIAA0424, PEM 2, PEM 2 homolog, PEM2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23229, 236915, 66013 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43307, Q3UTH8, Q9QX73 | |
| Arhgef9 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction