missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Arhgef9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557263
This item is not returnable.
View return policy
Description
Immunogen sequence: DEEGWFPASF VRLWVNQEDE VEEGPSDVQN GHLDPNSDCL CLGRPLQNRD QMRANVINEI MSTERHYIKH LKDIC Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 56%.
The protein encoded by this gene is a Rho-like GTPase that switches between the active state and inactive state to regulate CDC42 and other genes. Defects in this gene are a cause of startle disease with epilepsy, also known as hyperekplexia with epilepsy. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| Arhgef9 | |
| Polyclonal | |
| Unconjugated | |
| ARHGEF9 | |
| 9630036L12Rik; A230067K14; ARHDH9; ARHGEF9; Cdc42 guanine nucleotide exchange factor (GEF) 9; Cdc42 guanine nucleotide exchange factor 9; COLLYBISTIN; collybistin I; EIEE8; HPEM-2; hPEM-2 collybistin; KIAA0424; mKIAA0424; PEM2; PEM-2; PEM-2 homolog; rac/Cdc42 guanine nucleotide exchange factor 9; Rho guanine nucleotide exchange factor 9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23229 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43307 | |
| ARHGEF9 | |
| Recombinant protein corresponding to Human Arhgef9. Recombinant protein control fragment (Product #RP-95444). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction