missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGEF7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552097
This item is not returnable.
View return policy
Description
Immunogen sequence: RMSGFIYQGK LPTTGMTITK LEDSENHRNA FEISGSMIER ILVSCNNQQD LQEWVEHLQK QTKVTSVGNP TIKPHSVPSH TLPSHPVTPS SKHADSKPAP LTPAYHTLPH PSHHGTPHTT INWGPLEPPK TPKPW Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.
The serine/threonine kinase, p21 activated kinase (PAK), is a downstream effector of the small GTPases Cdc42 and Rac. PAK associates with Nck, the p85 and p110 subunits of PI3-kinase and PIX (PAK-interacting exchange factor) in a focal complex. The binding of PIX is necessary for the localization and activation of PAK in the Cdc42 to Rac signaling pathway, and this binding occurs through the high affinity of the N-terminal SH3 domain of PIX for a conserved proline rich PAK sequence. PIX exists as two isoforms, alpha and beta, and both are highly expressed in heart, muscle and thymus tissues of human and rat. alphaPIX is phosphorylated via PDGF and EphB2 receptor signaling pathways or through association with PI3-kinase. The alphaPIX isoform predominantly acts as a guanine nucleotide exchange factor (GEF) on Rac, which may mediate lamellipodia formation.
Specifications
| ARHGEF7 | |
| Polyclonal | |
| Unconjugated | |
| Arhgef7 | |
| ARHGEF7; betaPix; Beta-Pix; betaPix-b; betaPix-c; Cool; COOL1; cool-1; KIAA0142; mKIAA0142; Nbla10314; P50; P50BP; p85; p85Cool1; p85SPR; PAK3; Pak3 binding protein; Pak3bp; PAK-interacting exchange factor beta; PIX; Pixb; PIXbeta; Rho guanine nucleotide exchange factor (GEF) 7; Rho guanine nucleotide exchange factor (GEF7); Rho guanine nucleotide exchange factor 7; SH3 domain-containing proline-rich protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8874 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14155 | |
| Arhgef7 | |
| Recombinant protein corresponding to Human ARHGEF7. Recombinant protein control fragment (Product #RP-88620). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction