missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGEF7 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92602-0.1ml
This item is not returnable.
View return policy
Description
ARHGEF7 Polyclonal antibody specifically detects ARHGEF7 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ARHGEF7 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Beta-Pix, COOL1COOL-1, DKFZp686C12170, DKFZp761K1021, guanine nucleotide exchange factor 7, KIAA0142KIAA0412, Nbla10314, P50, P50BP, P85, P85COOL1, P85SPRBETA-PIX, PAK3, PAK3BP, PIXBPAK-interacting exchange factor beta, Rho guanine nucleotide exchange factor (GEF) 7, rho guanine nucleotide exchange factor 7, SH3 domain-containing proline-rich protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human ARHGEF7 (NP_001106984.1). MNSAEQTVTWLITLGVLESPKKTISDPEGFLQASLKDGVVLCRLLERLLPGTIEKTFDANDLYQGQNFNKVLSSLVTLNKVTADIGLGSDSVCARPSSHRIKSFDSLGSQSLHTRTSKLFQGQYRSLDMTDNSNNQLVVRAKFNFQQTNEDELSFSKGDVIHVTRVEEGGWWEGTLNGRTGWFPSNYVREVKASEKPVSPKSGTLKSPPKGFDTTAINKSYYNVVLQNILETENEYSKELQTVLSTYLRPLQTSEKLSSA | |
| 0.1 mL | |
| Cancer, Cell Biology, Cytoskeleton Markers, GPCR, Signal Transduction | |
| 8874 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction