missing translation for 'onlineSavingsMsg'
Learn More

ARHGEF6 Antibody, Novus Biologicals™

Product Code. 18357748 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18357748 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18357748 Supplier Novus Biologicals Supplier No. H00009459D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ARHGEF6 Polyclonal antibody specifically detects ARHGEF6 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGEF6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_004831.1
Gene Alias alphaPIX, Alpha-Pix, COOL-2, COOL2alpha, KIAA0006Cool2, mental retardation, X-linked 46, MRX46, PAK-interacting exchange factor alpha, PIXA, Rac/Cdc42 guanine exchange factor (GEF) 6, Rac/Cdc42 guanine nucleotide exchange factor (GEF) 6, Rac/Cdc42 guanine nucleotide exchange factor 6, rho guanine nucleotide exchange factor 6
Host Species Rabbit
Immunogen ARHGEF6 (NP_004831.1, 1 a.a. - 776 a.a.) full-length human protein. MNPEEQIVTWLISLGVLESPKKTICDPEEFLKSSLKNGVVLCKLINRLMPGSVEKFCLDPQTEADCINNINDFLKGCATLQVEIFDPDDLYSGVNFSKVLSTLLAVNKATEDQLSERPCGRSSSLSAANTSQTNPQGAVSSTVSGLQRQSKTVEMTENGSHQLIVKARFNFKQTNEDELSVCKGDIIYVTRVEEGGWWEGTLNGRTGWFPSNYVREIKSSERPLSPKAVKGFETAPLTKNYYTVVLQNILDTEKEYAKELQSLLVTYLRPLQSNNNLSTVEVTSLLGNFEEVCTFQQTLCQALEECSKFPENQHKVGGCLLSLMPHFKSMYLAYCANHPSAVNVLTQHSDELEQFMENQGASSPGILILTTNLSKPFMRLEKYVTLLQELERHMEDTHPDHQDILKAIVAFKTLMGQCQDLRKRKQLELQILSEPIQAWEGEDIKNLGNVIFMSQVMVQYGACEEKEERYLMLFSNVLIMLSASPRMSGFIYQGKIPIAGTVVTRLDEIEGNDCTFEITGNTVERIVVHCNNNQDFQEWLEQLNRLIRGPASCSSLSKTSSSSCSAHSSFSSTGQPRGPLEPPQIIKPWSLSCLRPAPPLRPSAALGYKERMSYILKESSKSPKTMKKFLHKRKTERKPSEEEYVIRKSTAALEEDAQILKVIEAYCTSANFQQGHGSSTRKDSIPQVLLPEEEKLIIEETRSNGQTIMEEKSLVDTVYALKDEVRELKQENKRMKQCLEEELKSRRDLEKLVRRLLKQTDECIRGESSSKTSILP
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9459
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.