missing translation for 'onlineSavingsMsg'
Learn More

ARHGEF6 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18408964 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408964 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408964 Supplier Novus Biologicals Supplier No. NBP130451FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ARHGEF6 Polyclonal antibody specifically detects ARHGEF6 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGEF6
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias alphaPIX, Alpha-Pix, COOL-2, COOL2alpha, KIAA0006Cool2, mental retardation, X-linked 46, MRX46, PAK-interacting exchange factor alpha, PIXA, Rac/Cdc42 guanine exchange factor (GEF) 6, Rac/Cdc42 guanine nucleotide exchange factor (GEF) 6, Rac/Cdc42 guanine nucleotide exchange factor 6, rho guanine nucleotide exchange factor 6
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 726 and 776 of human Rac/Cdc42 guanine nucleotide exchange factor 6 using the numbering given in entry NP_004831.1 (GeneID 9459).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9459
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.