missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGEF4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114876
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ARHGEF4.
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form complex with G proteins and stimulate Rho-dependent signals. This protein is similar to rat collybistin protein. Alternative splicing of this gene generates two transcript variants which encode different isoforms. Also there is possibility for the usage of multiple polyadenylation sites for this gene.
Specifications
| ARHGEF4 | |
| Polyclonal | |
| Unconjugated | |
| Arhgef4 | |
| 9330140K16Rik; APC-stimulated guanine nucleotide exchange factor; APC-stimulated guanine nucleotide exchange factor 1; Arhgef4; Asef; ASEF1; DKFZp434G2016; ENSMUSG00000070955; GEF4; Kiaa1112; LOC100067370; LOW QUALITY PROTEIN: rho guanine nucleotide exchange factor 4; Rho guanine nucleotide exchange factor (GEF) 4; Rho guanine nucleotide exchange factor 4; STM6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 226970, 301334, 50649 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q7TNR9, Q9NR80 | |
| Arhgef4 | |
| A synthesized peptide derived from human ARHGEF4(Accession Q9NR80), corresponding to amino acid residues R327-E377. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction