missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGEF4 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-1316
This item is not returnable.
View return policy
Description
ARHGEF4 Polyclonal specifically detects ARHGEF4 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| ARHGEF4 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| APC-stimulated guanine nucleotide exchange factor 1, Asef, ASEF1, ASEFAPC-stimulated guanine nucleotide exchange factor, GEF4, KIAA1112, Rho guanine nucleotide exchange factor (GEF) 4, rho guanine nucleotide exchange factor 4, STM6 | |
| Goat | |
| 79 kDa | |
| 0.1 mg | |
| Signal Transduction | |
| 50649 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 2 - 4 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| Q9NR80 | |
| ARHGEF4 | |
| Peptide with sequence PWEEPAGEKPSCS corresponding to N-Terminus according to NP_056135.2, NP_127462.1. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_056135.2; NP_127462.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction